Gene Transcripts

Gene Symbol
OR2A1
Entrez Gene ID
346528
Gene Name
olfactory receptor family 2 subfamily A member 1
Gene Aliases
Chromosome Location
Chr. 7: 144318125 - 144319057 on Build GRCh38
UniGene ID
Hs.528398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005287.1 NP_001005287.1 1 103

Gene Symbol
OR2A42
Entrez Gene ID
402317
Gene Name
olfactory receptor family 2 subfamily A member 42
Gene Aliases
Chromosome Location
Chr. 7: 144231911 - 144232843 on Build GRCh38
UniGene ID
Hs.591830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001802.2 NP_001001802.2 1 103

View Full Product Details