Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024334.2 NP_077310.1 6 710
XM_017007176.1 XP_016862665.1 7 832
GenBank mRNA AK027757.1 - 6 636
AK027877.1 - 6 574
AK074073.1 - 4 246
AK075010.1 - 6 561
AK225589.1 - 6 633
AL136916.1 - 6 604
AY358625.1 - 6 473
BC003125.1 - 6 605 3503636
BC008054.2 - 4 200
BC011719.2 - 6 605 4079766
KU178769.1 - 5 456
KU178770.1 - 5 456

View Full Product Details