Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127394.3 | NP_001120866.1 | 4 | 1193 | |
NM_001300764.1 | NP_001287693.1 | 5 | 1347 | ||
NM_001300766.1 | NP_001287695.1 | 5 | 1231 | ||
NM_052965.3 | NP_443197.1 | 5 | 1335 | ||
NR_023349.2 | 6 | 1452 | |||
NR_125335.1 | 6 | 1415 | |||
GenBank mRNA | AF288394.1 | - | 5 | 1310 | |
AK098196.1 | - | 5 | 1233 | ||
AK296655.1 | - | 4 | 1088 | ||
AK307241.1 | - | 6 | 1343 | ||
BC022030.1 | - | 5 | 1280 | 4824025 | |
BM789892.1 | - | 1 | 56 |