Gene Transcripts

Gene Symbol
Obox3
Entrez Gene ID
246791
Gene Name
oocyte specific homeobox 3
Gene Aliases
Ohx
Chromosome Location
Chr. 7: 15625306 - 15639777 on Build GRCm38
UniGene ID
Mm.425362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145707.3 NP_663753.3 4 404
GenBank mRNA AK139750.1 - 4 405
AY094981.1 - 794
BC119564.1 - 1 792

View Full Product Details