Gene Transcripts

Gene Symbol
Mup7
Entrez Gene ID
100041658
Gene Name
major urinary protein 7
Gene Aliases
Gm12546, Mup3, OTTMUSG00000007428
Chromosome Location
Chr. 4: 60066469 - 60070481 on Build GRCm38
UniGene ID
Mm.457982
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134675.1 NP_001128147.1 7 970

Gene Symbol
Mup1
Entrez Gene ID
17840
Gene Name
major urinary protein 1
Gene Aliases
Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
Chromosome Location
Chr. 4: 60498012 - 60501960 on Build GRCm38
UniGene ID
Mm.335875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC091744.1 - 802
BC092254.1 - 799 5100458
BC100586.1 - 819 6769126

Gene Symbol
Mup15
Entrez Gene ID
100039150
Gene Name
major urinary protein 15
Gene Aliases
Gm2068, Mup13
Chromosome Location
Chr. 4: 61435787 - 61439714 on Build GRCm38
UniGene ID
Mm.237772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200004.1 NP_001186933.1 7 826

View Full Product Details