Gene Transcripts
- Gene Symbol
- Vmn2r21
- Entrez Gene ID
- 546912
- Gene Name
- vomeronasal 2, receptor 21
- Gene Aliases
- EG546912
- Chromosome Location
- Chr. 6: 123492508 - 123533406 on Build GRCm38
- UniGene ID
- Mm.469876
- Species
- Mus musculus
- Gene Symbol
- Vmn2r20
- Entrez Gene ID
- 667180
- Gene Name
- vomeronasal 2, receptor 20
- Gene Aliases
- EG667180
- Chromosome Location
- Chr. 6: 123385262 - 123418061 on Build GRCm38
- UniGene ID
- Mm.469877
- Species
- Mus musculus
- Gene Symbol
- Vmn2r22
- Entrez Gene ID
- 546913
- Gene Name
- vomeronasal 2, receptor 22
- Gene Aliases
- EG546913
- Chromosome Location
- Chr. 6: 123609758 - 123650635 on Build GRCm38
- UniGene ID
- Mm.469875
- Species
- Mus musculus
View Full Product Details