Gene Transcripts

Gene Symbol
Vmn2r29
Entrez Gene ID
76229
Gene Name
vomeronasal 2, receptor 29
Gene Aliases
6430701C03Rik
Chromosome Location
Chr. 7: 7212996 - 7278289 on Build GRCm38
UniGene ID
Mm.387947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113468.1 NP_001106939.1 3 1027

View Full Product Details