Gene Transcripts

Gene Symbol
Srpx
Entrez Gene ID
51795
Gene Name
sushi-repeat-containing protein
Gene Aliases
drs, drs-1, drs-2, mDRS-1, mDRS-2
Chromosome Location
Chr. X: 10037977 - 10117661 on Build GRCm38
UniGene ID
Mm.1927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016911.4 NP_058607.1 2, 3 271
GenBank mRNA AB028049.1 - 2, 3 198
AK034696.1 - 2, 3 273
BC022572.1 - 2, 3 227 3993025

View Full Product Details