Gene Transcripts

Gene Symbol
Olr4
Entrez Gene ID
292560
Gene Name
olfactory receptor 4
Gene Aliases
ratchr1-65841497-65842435_ORF
Chromosome Location
Chr. 1: 70454322 - 70455260 on Build Rnor_6.0
UniGene ID
Rn.142005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000111.1 NP_001000111.1 1 334

View Full Product Details