Gene Transcripts

Gene Symbol
Vom2r23
Entrez Gene ID
308303
Gene Name
vomeronasal 2 receptor, 23
Gene Aliases
RGD1560836
Chromosome Location
Chr. 8: 4019722 - 4050517 on Build Rnor_6.0
UniGene ID
Rn.217632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099649.1 NP_001093119.1 1 104

View Full Product Details