Gene Transcripts
- Gene Symbol
- NBPF14
- Entrez Gene ID
- 25832
- Gene Name
- neuroblastoma breakpoint family member 14
- Gene Aliases
- DJ328E19.C1.1, NBPF
- Chromosome Location
- Chr. 1: 148531385 - 148607636 on Build GRCh38
- UniGene ID
- Hs.534675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL117237.1 |
- |
10 |
1136 |
|
- Gene Symbol
- NBPF26
- Entrez Gene ID
- 101060684
- Gene Name
- neuroblastoma breakpoint family member 26
- Gene Aliases
- Chromosome Location
- Chr. 1: 120805561 - 120842215 on Build GRCh38
- UniGene ID
- Hs.534675
- Species
- Homo sapiens
View Full Product Details