Gene Transcripts
- Gene Symbol
- NIPBL
- Entrez Gene ID
- 25836
- Gene Name
- NIPBL, cohesin loading factor
- Gene Aliases
- CDLS, CDLS1, IDN3, IDN3-B, Scc2
- Chromosome Location
- Chr. 5: 36876759 - 37065819 on Build GRCh38
- UniGene ID
- Hs.481927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015384.4 |
NP_056199.2
|
25, 26 |
5510 |
|
NM_133433.3 |
NP_597677.2
|
25, 26 |
5510 |
|
XM_005248280.3 |
XP_005248337.1
|
25, 26 |
5499 |
|
XM_005248282.4 |
XP_005248339.3
|
19, 20 |
5001 |
|
XM_006714467.2 |
XP_006714530.1
|
25, 26 |
5499 |
|
XM_006714468.2 |
XP_006714531.1
|
24, 25 |
5301 |
|
XM_011514015.1 |
XP_011512317.1
|
25, 26 |
5499 |
|
XM_017009329.1 |
XP_016864818.1
|
25, 26 |
5499 |
|
XM_017009330.1 |
XP_016864819.1
|
17, 18 |
7773 |
|
XM_017009331.1 |
XP_016864820.1
|
24, 25 |
3873 |
|
GenBank mRNA |
AJ627032.1 |
- |
25, 26 |
5479 |
|
AJ640137.1 |
- |
25, 26 |
5479 |
|
KJ807791.1 |
- |
11, 12 |
1297 |
|
View Full Product Details