Gene Transcripts

Gene Symbol
RANBP6
Entrez Gene ID
26953
Gene Name
RAN binding protein 6
Gene Aliases
Chromosome Location
Chr. 9: 6011019 - 6015660 on Build GRCh38
UniGene ID
Hs.167496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012416.3 NP_036548.1 1 994
GenBank mRNA AF039023.1 - 1 963
AK307840.1 - 1 969
BC098406.1 - 1 969 30347107
BX537405.1 - 1 973

View Full Product Details