Gene Transcripts

Gene Symbol
MRPS16
Entrez Gene ID
51021
Gene Name
mitochondrial ribosomal protein S16
Gene Aliases
CGI-132, COXPD2, MRP-S16, RPMS16
Chromosome Location
Chr. 10: 73248843 - 73252693 on Build GRCh38
UniGene ID
Hs.180312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016065.3 NP_057149.1 3 565
GenBank mRNA AB049948.1 - 3 451
AF151890.1 - 3 518
AK025715.1 - 3 498
AK098291.1 - 3 511
BC021106.1 - 3 460 5020046

View Full Product Details