Gene Transcripts

Gene Symbol
NDUFA13
Entrez Gene ID
51079
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
Gene Aliases
B16.6, CDA016, CGI-39, GRIM-19, GRIM19
Chromosome Location
Chr. 19: 19516210 - 19528204 on Build GRCh38
UniGene ID
Hs.534453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015965.6 NP_057049.5 11
GenBank mRNA AF261134.1 - 1 442
BG724220.1 - 1 486
BM839920.1 - 11

View Full Product Details