Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016125.3 | NP_057209.3 | 6 | 1082 | |
GenBank mRNA | AK098649.1 | - | 4 | 462 | |
AK296197.1 | - | 4 | 885 | ||
BC006971.1 | - | 5 | 959 | ||
BC088365.1 | - | 6 | 1045 | 30390062 |