Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278710.1 | NP_001265639.1 | 4 | 464 | |
NM_001278711.1 | NP_001265640.1 | 4 | 703 | ||
NM_016588.2 | NP_057672.1 | 3 | 962 | ||
XM_006715106.3 | XP_006715169.1 | 5 | 1353 | ||
XM_006715107.3 | XP_006715170.1 | 3 | 1143 | ||
GenBank mRNA | AF114833.1 | - | 3 | 493 | |
AF136631.1 | - | 3 | 479 | ||
AJ420483.1 | - | 3 | 436 | ||
AK093824.1 | - | 3 | 1157 | ||
AK314095.1 | - | 3 | 515 | ||
AL541038.3 | - | 3 | 547 | ||
BC002683.2 | - | 3 | 440 | 3605775 | |
BC042019.1 | - | 3 | 963 | 5312094 | |
BF970432.1 | - | 3 | 355 | ||
BX340783.2 | - | 4 | 699 | ||
DB013243.1 | - | 4 | 464 |