Gene Transcripts
- Gene Symbol
- MRPL39
- Entrez Gene ID
- 54148
- Gene Name
- mitochondrial ribosomal protein L39
- Gene Aliases
- C21orf92, L39mt, MRP-L5, MRPL5, MSTP003, PRED22, PRED66, RPML5
- Chromosome Location
- Chr. 21: 25585656 - 25607858 on Build GRCh38
- UniGene ID
- Hs.420696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017446.3 |
NP_059142.2
|
6, 7 |
720 |
|
NM_080794.3 |
NP_542984.2
|
6, 7 |
720 |
|
XM_006724026.3 |
XP_006724089.1
|
6, 7 |
712 |
|
XM_011529651.2 |
XP_011527953.1
|
6, 7 |
791 |
|
GenBank mRNA |
AF109357.1 |
- |
6, 7 |
694 |
|
AF239727.1 |
- |
6, 7 |
706 |
|
AF270511.1 |
- |
3, 4 |
259 |
|
AK000458.1 |
- |
6, 7 |
711 |
|
AK225878.1 |
- |
6, 7 |
711 |
|
BC004896.2 |
- |
6, 7 |
693 |
|
BC107719.1 |
- |
6, 7 |
716 |
6737868 |
View Full Product Details