Gene Transcripts
- Gene Symbol
- LRRC36
- Entrez Gene ID
- 55282
- Gene Name
- leucine rich repeat containing 36
- Gene Aliases
- RORBP70, XLHSRF2
- Chromosome Location
- Chr. 16: 67326844 - 67385206 on Build GRCh38
- UniGene ID
- Hs.125139
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161575.1 |
NP_001155047.1
|
8, 9 |
1548 |
|
NM_018296.5 |
NP_060766.5
|
11, 12 |
1818 |
|
XM_005256025.2 |
XP_005256082.1
|
11, 12 |
1818 |
|
XM_005256026.2 |
XP_005256083.1
|
10, 11 |
1377 |
|
XM_005256028.1 |
XP_005256085.1
|
10, 11 |
1690 |
|
XM_011523201.1 |
XP_011521503.1
|
8, 9 |
1438 |
|
XM_011523202.2 |
XP_011521504.1
|
7, 8 |
1293 |
|
XM_011523203.1 |
XP_011521505.1
|
8, 9 |
1200 |
|
XM_011523204.1 |
XP_011521506.1
|
9, 10 |
1617 |
|
XM_011523205.1 |
XP_011521507.1
|
10, 11 |
1708 |
|
XM_017023400.1 |
XP_016878889.1
|
11, 12 |
1818 |
|
GenBank mRNA |
AK292311.1 |
- |
8, 9 |
1527 |
|
AY313780.1 |
- |
8, 9 |
1511 |
|
AY963267.1 |
- |
11, 12 |
1818 |
|
BC018031.1 |
- |
10, 11 |
1312 |
|
BC026156.2 |
- |
11, 12 |
1818 |
4828487 |
View Full Product Details