Gene Transcripts
- Gene Symbol
- OGDHL
- Entrez Gene ID
- 55753
- Gene Name
- oxoglutarate dehydrogenase-like
- Gene Aliases
- Chromosome Location
- Chr. 10: 49734641 - 49762379 on Build GRCh38
- UniGene ID
- Hs.17860
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143996.1 |
NP_001137468.1
|
18, 19 |
2498 |
|
NM_001143997.1 |
NP_001137469.1
|
17, 18 |
2293 |
|
NM_018245.2 |
NP_060715.2
|
19, 20 |
2669 |
|
XM_011539946.1 |
XP_011538248.1
|
19, 20 |
2554 |
|
XM_011539947.1 |
XP_011538249.1
|
19, 20 |
2760 |
|
XM_017016401.1 |
XP_016871890.1
|
19, 20 |
2944 |
|
XM_017016402.1 |
XP_016871891.1
|
19, 20 |
2571 |
|
XM_017016403.1 |
XP_016871892.1
|
17, 18 |
2317 |
|
GenBank mRNA |
AB033116.3 |
- |
18, 19 |
2532 |
|
AK001713.1 |
- |
19, 20 |
2613 |
|
AK055817.1 |
- |
1, 2 |
1563 |
|
AK290226.1 |
- |
18, 19 |
2642 |
|
AK296551.1 |
- |
18, 19 |
2407 |
|
AK303729.1 |
- |
17, 18 |
2240 |
|
BC026320.1 |
- |
19, 20 |
2588 |
4797814 |
View Full Product Details