Gene Transcripts
- Gene Symbol
- PHRF1
- Entrez Gene ID
- 57661
- Gene Name
- PHD and ring finger domains 1
- Gene Aliases
- PPP1R125, RNF221
- Chromosome Location
- Chr. 11: 576446 - 612225 on Build GRCh38
- UniGene ID
- Hs.325838
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286581.1 |
NP_001273510.1
|
13, 14 |
1788 |
|
NM_001286582.1 |
NP_001273511.1
|
13, 14 |
1782 |
|
NM_001286583.1 |
NP_001273512.1
|
13, 14 |
1776 |
|
NM_020901.3 |
NP_065952.2
|
13, 14 |
1785 |
|
XM_005253025.4 |
XP_005253082.1
|
13, 14 |
1746 |
|
XM_005253027.3 |
XP_005253084.1
|
13, 14 |
1734 |
|
XM_011520236.2 |
XP_011518538.1
|
13, 14 |
1782 |
|
XM_011520237.2 |
XP_011518539.1
|
12, 13 |
1629 |
|
GenBank mRNA |
AB040975.1 |
- |
13, 14 |
1748 |
|
BC112931.1 |
- |
11, 12 |
3530 |
|
BC136615.1 |
- |
13, 14 |
1713 |
9020605 |
BC144293.1 |
- |
13, 14 |
1710 |
9052813 |
BC144294.1 |
- |
13, 14 |
1701 |
9052814 |
BC144295.1 |
- |
13, 14 |
1707 |
9052815 |
BC146771.1 |
- |
13, 14 |
1748 |
9007218 |
View Full Product Details