Gene Transcripts
- Gene Symbol
- MUC3A
- Entrez Gene ID
- 4584
- Gene Name
- mucin 3A, cell surface associated
- Gene Aliases
- MUC-3A, MUC3
- Chromosome Location
- Chr. 7: 100949054 - 100968347 on Build GRCh38
- UniGene ID
- Hs.703577
- Species
- Homo sapiens
- Gene Symbol
- MUC3B
- Entrez Gene ID
- 57876
- Gene Name
- mucin 3B, cell surface associated
- Gene Aliases
- MUC3, MUC3A
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB038783.1 |
- |
9 |
2472 |
|
View Full Product Details