Gene Transcripts
- Gene Symbol
- OSTC
- Entrez Gene ID
- 58505
- Gene Name
- oligosaccharyltransferase complex non-catalytic subunit
- Gene Aliases
- DC2
- Chromosome Location
- Chr. 4: 108650585 - 108667822 on Build GRCh38
- UniGene ID
- Hs.445803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267818.1 |
NP_001254747.1
|
2, 3 |
314 |
|
NM_021227.3 |
NP_067050.1
|
2, 3 |
314 |
|
GenBank mRNA |
AF068297.1 |
- |
2, 3 |
277 |
|
AF161425.1 |
- |
2, 3 |
298 |
|
AF201937.1 |
- |
2, 3 |
305 |
|
AF343342.1 |
- |
2, 3 |
243 |
|
AK312185.1 |
- |
2, 3 |
258 |
|
BC016321.1 |
- |
2, 3 |
302 |
4075646 |
BC054857.1 |
- |
2, 3 |
304 |
4093030 |
BM471860.1 |
- |
|
242 |
|
BM541430.1 |
- |
2, 3 |
265 |
|
BM546028.1 |
- |
2, 3 |
304 |
|
BQ435510.1 |
- |
|
314 |
|
CN430159.1 |
- |
2, 3 |
252 |
|
View Full Product Details