Gene Transcripts

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
Chromosome Location
Chr. 2: 111766147 - 111884196 on Build GRCh38
UniGene ID
Hs.436527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022662.3 NP_073153.1 10, 11 1502
XM_011511633.2 XP_011509935.1 10, 11 1534
XM_011511634.2 XP_011509936.1 10, 11 1649
XM_011511636.2 XP_011509938.1 10, 11 1534
XM_011511637.2 XP_011509939.1 10, 11 1534
XM_017004710.1 XP_016860199.1 10, 11 1534
XM_017004711.1 XP_016860200.1 10, 11 1534
XM_017004712.1 XP_016860201.1 10, 11 1649
XM_017004713.1 XP_016860202.1 9, 10 1597
XM_017004714.1 XP_016860203.1 9, 10 1545
XM_017004715.1 XP_016860204.1 10, 11 1534
GenBank mRNA AJ278357.1 - 10, 11 1517
BC104902.1 - 10, 11 1304 8143905
BC104904.1 - 10, 11 1304 8143907

View Full Product Details