Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304833.1 | NP_001291762.1 | 7 | 1465 | |
NM_001304834.1 | NP_001291763.1 | 8 | 1896 | ||
NM_001304835.1 | NP_001291764.1 | 7 | 2016 | ||
NM_001304836.1 | NP_001291765.1 | 7 | 1785 | ||
NM_001304837.1 | NP_001291766.1 | 6 | 1646 | ||
NM_001304838.1 | NP_001291767.1 | 7 | 1703 | ||
NM_024623.2 | NP_078899.1 | 8 | 1852 | ||
GenBank mRNA | AK023553.1 | - | 8 | 1748 | |
AK094820.1 | - | 7 | 1448 | ||
AK095481.1 | - | 6 | 1554 | ||
BC098119.1 | - | 8 | 1628 | 40024912 | |
BC098293.1 | - | 8 | 1628 | 40024914 | |
BC101940.1 | - | 7 | 1435 | 40024915 | |
BC105637.1 | - | 7 | 1435 | 40024913 | |
BX640734.1 | - | 8 | 1496 |