Gene Transcripts

Gene Symbol
LRRK1
Entrez Gene ID
79705
Gene Name
leucine rich repeat kinase 1
Gene Aliases
RIPK6, Roco1
Chromosome Location
Chr. 15: 100919017 - 101070837 on Build GRCh38
UniGene ID
Hs.407918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024652.4 NP_078928.3 27 4570
XM_005254979.3 XP_005255036.1 13 2228
XM_011522012.2 XP_011520314.1 29 4844
XM_011522013.2 XP_011520315.1 27 4553
XM_011522014.2 XP_011520316.1 27 4768
XM_011522015.2 XP_011520317.1 22 5363
XM_011522016.2 XP_011520318.1 25 4045
XM_011522017.2 XP_011520319.1 13 2235
XM_011522018.2 XP_011520320.1 13 2230
XM_017022570.1 XP_016878059.1 17 2817
GenBank mRNA AB058693.1 - 13 2275
AK054662.1 - 1 18
AK058028.1 - 7 1118
AK131075.1 - 8 1503
BC005408.1 - 2 241
BC020379.1 - 2 241
BC144705.1 - 25 3958
BC146769.1 - 13 2275
BC171802.1 - 26 4244
DQ013130.1 - 26 4208

View Full Product Details