Gene Transcripts

Gene Symbol
NUAK2
Entrez Gene ID
81788
Gene Name
NUAK family kinase 2
Gene Aliases
SNARK
Chromosome Location
Chr. 1: 205302059 - 205321791 on Build GRCh38
UniGene ID
Hs.497512
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030952.2 NP_112214.2 2 492
GenBank mRNA AK074830.1 - 2 446
AK303577.1 - 2 492
AL136891.1 - 2 456
BC017306.2 - 2 428 5088037

View Full Product Details