Gene Transcripts

Gene Symbol
VANGL1
Entrez Gene ID
81839
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
KITENIN, LPP2, STB2, STBM2
Chromosome Location
Chr. 1: 115641953 - 115698224 on Build GRCh38
UniGene ID
Hs.515130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172411.1 NP_001165882.1 2 315
NM_001172412.1 NP_001165883.1 2 259
NM_138959.2 NP_620409.1 2 315
GenBank mRNA AB057596.1 - 2 259
AB075805.1 - 1 56
AF481859.1 - 2 277
BC065272.1 - 2 292 6165098
BM552933.1 - 334

View Full Product Details