Gene Transcripts

Gene Symbol
PARP10
Entrez Gene ID
84875
Gene Name
poly(ADP-ribose) polymerase family member 10
Gene Aliases
ARTD10
Chromosome Location
Chr. 8: 143977152 - 143986471 on Build GRCh38
UniGene ID
Hs.348609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317895.1 NP_001304824.1 3 813
NM_032789.4 NP_116178.2 4 689
NR_134234.1 4 689
XM_011517336.2 XP_011515638.1 4 656
GenBank mRNA AK027370.1 - 4 646
AK128537.1 - 5 504
AK222914.1 - 4 672
AK303317.1 - 3 738
AK308657.1 - 3 493
AK309251.1 - 3 393
BC020378.1 - 4 664
BC030549.1 - 4 663
BC132961.1 - 4 640 40146983
BC136542.1 - 4 640 9020532
BC144235.1 - 4 640

View Full Product Details