Gene Transcripts

Gene Symbol
LYRM7
Entrez Gene ID
90624
Gene Name
LYR motif containing 7
Gene Aliases
C5orf31, MC3DN8, MZM1L
Chromosome Location
Chr. 5: 131170914 - 131205426 on Build GRCh38
UniGene ID
Hs.115467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181705.3 NP_859056.2 4 296
GenBank mRNA AK000803.1 - 4 267
BC037930.1 - 4 248 5284728
BI601105.1 - 4 272
DA996312.1 - 4 298

View Full Product Details