Gene Transcripts

Gene Symbol
SLC51B
Entrez Gene ID
123264
Gene Name
solute carrier family 51 beta subunit
Gene Aliases
OSTB, OSTBETA
Chromosome Location
Chr. 15: 65045370 - 65053396 on Build GRCh38
UniGene ID
Hs.27018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178859.3 NP_849190.2 3, 4 502
XM_005254159.4 XP_005254216.1 3, 4 477
GenBank mRNA AK129601.1 - 3, 4 405
AY194242.1 - 2, 3 211
BC103842.1 - 2, 3 208 40002450
BC103843.1 - 2, 3 208 40002452
BC103844.1 - 2, 3 208 40002453

View Full Product Details