Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133452.2 | NP_597709.2 | 2 | 378 | |
XM_011527671.1 | XP_011525973.1 | 2 | 372 | ||
GenBank mRNA | BC037428.1 | - | 2 | 270 | |
BX401103.2 | - | 2 | 274 | ||
DB446023.1 | - | 2 | 380 |