Gene Transcripts

Gene Symbol
TMEM252
Entrez Gene ID
169693
Gene Name
transmembrane protein 252
Gene Aliases
C9orf71
Chromosome Location
Chr. 9: 68536580 - 68540867 on Build GRCh38
UniGene ID
Hs.663056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153237.1 NP_694969.1 2 345
GenBank mRNA BC029780.1 - 2 345

View Full Product Details