Gene Transcripts
- Gene Symbol
- OR8U1
- Entrez Gene ID
- 219417
- Gene Name
- olfactory receptor family 8 subfamily U member 1
- Gene Aliases
- Chromosome Location
- Chr. 11: 56375624 - 56376553 on Build GRCh38
- UniGene ID
- Hs.553618
- Species
- Homo sapiens
- Gene Symbol
- OR8U8
- Entrez Gene ID
- 504189
- Gene Name
- olfactory receptor family 8 subfamily U member 8
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
- Gene Symbol
- OR8U9
- Entrez Gene ID
- 504190
- Gene Name
- olfactory receptor family 8 subfamily U member 9
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.553618
- Species
- Homo sapiens
View Full Product Details