Gene Transcripts

Gene Symbol
OR8G5
Entrez Gene ID
219865
Gene Name
olfactory receptor family 8 subfamily G member 5
Gene Aliases
OR11-298, OR8G5P, OR8G6
Chromosome Location
Chr. 11: 124241158 - 124323283 on Build GRCh38
UniGene ID
Hs.722725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005198.1 NP_001005198.1 1 42
GenBank mRNA BC136820.1 - 1 65
BC136822.1 - 1 65 9020813

View Full Product Details