Gene Transcripts

Gene Symbol
NAALADL2
Entrez Gene ID
254827
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
Chromosome Location
Chr. 3: 174439669 - 175809510 on Build GRCh38
UniGene ID
Hs.565848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207015.2 NP_996898.2 2 511
XM_006713560.2 XP_006713623.1 4 594
XM_011512612.2 XP_011510914.1 2 3848
XM_011512613.1 XP_011510915.1 2 656
XM_011512615.2 XP_011510917.1 2 3849
XM_011512617.2 XP_011510919.1 2 3846
XM_017006070.1 XP_016861559.1 6 1036
XM_017006071.1 XP_016861560.1 7 1106
XM_017006072.1 XP_016861561.1 8 1240
XM_017006073.1 XP_016861562.1 6 839
XM_017006074.1 XP_016861563.1 8 3258
XM_017006075.1 XP_016861564.1 5 1983
XM_017006076.1 XP_016861565.1 5 776
XM_017006077.1 XP_016861566.1 7 2187
XM_017006078.1 XP_016861567.1 5 2004
XM_017006079.1 XP_016861568.1 3 1480
XM_017006080.1 XP_016861569.1 3 1851
XM_017006081.1 XP_016861570.1 2 3848
XM_017006082.1 XP_016861571.1 5 1990
GenBank mRNA AJ607396.1 - 2 552
AK308416.1 - 2 527
AL832144.1 - 2 511
BC057243.1 - 2 514

View Full Product Details