Gene Transcripts
- Gene Symbol
- C1orf101
- Entrez Gene ID
- 257044
- Gene Name
- chromosome 1 open reading frame 101
- Gene Aliases
- Chromosome Location
- Chr. 1: 244452014 - 244640362 on Build GRCh38
- UniGene ID
- Hs.459534
- Species
- Homo sapiens
- Gene Symbol
- AASDHPPT
- Entrez Gene ID
- 60496
- Gene Name
- aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
- Gene Aliases
- AASD-PPT, ACPS, CGI-80, LYS2, LYS5
- Chromosome Location
- Chr. 11: 106077565 - 106098692 on Build GRCh38
- UniGene ID
- Hs.524009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537665.1 |
- |
1 |
2400 |
|
View Full Product Details