Gene Transcripts

Gene Symbol
LY6G6F
Entrez Gene ID
259215
Gene Name
lymphocyte antigen 6 complex, locus G6F
Gene Aliases
C6orf21, G6f, LY6G6D, NG32
Chromosome Location
Chr. 6: 31706907 - 31710595 on Build GRCh38
UniGene ID
Hs.591794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003693.1 NP_001003693.1 3 610
GenBank mRNA AJ496460.1 - 3 610
BC137212.1 - 3 654 9021209
BC137213.1 - 3 654 9021210

View Full Product Details