Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271156.2 | NP_001258085.2 | 2 | 1555 | |
NM_173832.5 | NP_776193.2 | 2 | 841 | ||
GenBank mRNA | AK096024.1 | - | 2 | 830 | |
AL831951.1 | - | 2 | 1266 | ||
BC034608.1 | - | 2 | 1555 | 4827717 | |
CR992458.1 | - | 1 | 287 | ||
DA216172.1 | - | 1 | 485 |