Gene Transcripts

Gene Symbol
OR2AG2
Entrez Gene ID
338755
Gene Name
olfactory receptor family 2 subfamily AG member 2
Gene Aliases
OR11-76, OR2AG2P
Chromosome Location
Chr. 11: 6768007 - 6768957 on Build GRCh38
UniGene ID
Hs.534614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004490.1 NP_001004490.1 1 480
GenBank mRNA BC140734.1 - 1 578

View Full Product Details