Gene Transcripts

Gene Symbol
LINGO4
Entrez Gene ID
339398
Gene Name
leucine rich repeat and Ig domain containing 4
Gene Aliases
DAAT9248, LRRN6D, PRO34002
Chromosome Location
Chr. 1: 151800264 - 151805442 on Build GRCh38
UniGene ID
Hs.591477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004432.3 NP_001004432.1 2 1610
GenBank mRNA AY358115.1 - 2 1577
BC137220.1 - 2 1434 9021217
BC137221.1 - 2 1434 9021218

View Full Product Details