Gene Transcripts

Gene Symbol
NRBP2
Entrez Gene ID
340371
Gene Name
nuclear receptor binding protein 2
Gene Aliases
TRG16, pp9320
Chromosome Location
Chr. 8: 143829776 - 143840974 on Build GRCh38
UniGene ID
Hs.521926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178564.3 NP_848659.2 13, 14 1275
XM_017013378.1 XP_016868867.1 13, 14 1275
XM_017013379.1 XP_016868868.1 13, 14 1275
XM_017013380.1 XP_016868869.1 13, 14 1275
XM_017013381.1 XP_016868870.1 13, 14 1275
XM_017013382.1 XP_016868871.1 13, 14 1275
XM_017013383.1 XP_016868872.1 13, 14 1275
GenBank mRNA AF318376.1 - 3, 4 615
AK293943.1 - 13, 14 1275
AL137662.1 - 11, 12 1204
AL834530.1 - 6, 7 514
AY277601.1 - 3, 4 602
BC071605.1 - 12, 13 1438
BC113873.1 - 10, 11 1279 40053695
BC114481.1 - 11, 12 1176
CR457350.1 - 5, 6 406

View Full Product Details