Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146339.1 | NP_001139811.1 | 2 | 128 | |
XM_011526902.2 | XP_011525204.1 | 3 | 748 | ||
XM_011526903.2 | XP_011525205.1 | 3 | 748 | ||
XM_011526904.2 | XP_011525206.1 | 2 | 273 |