Gene Transcripts
- Gene Symbol
- PQBP1
- Entrez Gene ID
- 10084
- Gene Name
- polyglutamine binding protein 1
- Gene Aliases
- MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
- Chromosome Location
- Chr. X: 48897862 - 48903145 on Build GRCh38
- UniGene ID
- Hs.534384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001032381.1 |
NP_001027553.1
|
4, 5 |
410 |
|
NM_001032382.1 |
NP_001027554.1
|
4, 5 |
478 |
|
NM_001032383.1 |
NP_001027555.1
|
4, 5 |
489 |
|
NM_001032384.1 |
NP_001027556.1
|
4, 5 |
398 |
|
NM_001167989.1 |
NP_001161461.1
|
4, 5 |
410 |
|
NM_001167990.1 |
NP_001161462.1
|
4, 5 |
368 |
|
NM_005710.2 |
NP_005701.1
|
3, 4 |
543 |
|
XM_005272571.3 |
XP_005272628.1
|
4, 5 |
473 |
|
XM_011543884.2 |
XP_011542186.1
|
4, 5 |
548 |
|
XM_017029207.1 |
XP_016884696.1
|
4, 5 |
539 |
|
GenBank mRNA |
AB016533.1 |
- |
4, 5 |
459 |
|
AB041834.1 |
- |
3, 4 |
289 |
|
AB041835.1 |
- |
3, 4 |
289 |
|
AJ005893.1 |
- |
3, 4 |
546 |
|
AJ242829.1 |
- |
4, 5 |
349 |
|
AJ973594.1 |
- |
3, 4 |
304 |
|
AJ973595.1 |
- |
3, 4 |
304 |
|
AJ973596.1 |
- |
|
304 |
|
AJ973597.1 |
- |
3, 4 |
304 |
|
AJ973598.1 |
- |
3, 4 |
304 |
|
AJ973599.1 |
- |
3, 4 |
304 |
|
AJ973600.1 |
- |
3, 4 |
304 |
|
AJ973601.1 |
- |
4, 5 |
391 |
|
AJ973602.1 |
- |
3, 4 |
608 |
|
AJ973603.1 |
- |
4, 5 |
391 |
|
AJ973604.1 |
- |
3, 4 |
308 |
|
AJ973606.1 |
- |
3, 4 |
304 |
|
BC012358.1 |
- |
4, 5 |
370 |
4662649 |
BE385548.1 |
- |
2, 3 |
224 |
|
BE396796.1 |
- |
4, 5 |
390 |
|
BI818141.1 |
- |
4, 5 |
372 |
|
BX362311.2 |
- |
4, 5 |
447 |
|
BX369697.2 |
- |
4, 5 |
630 |
|
CD051018.1 |
- |
4, 5 |
389 |
|
CN292761.1 |
- |
4, 5 |
478 |
|
CN292769.1 |
- |
4, 5 |
396 |
|
KU178500.1 |
- |
3, 4 |
289 |
|
KU178501.1 |
- |
4, 5 |
337 |
|
View Full Product Details