Gene Transcripts

Gene Symbol
OR2T33
Entrez Gene ID
391195
Gene Name
olfactory receptor family 2 subfamily T member 33
Gene Aliases
OR1-56
Chromosome Location
Chr. 1: 248272852 - 248273814 on Build GRCh38
UniGene ID
Hs.553783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004695.1 NP_001004695.1 1 773
GenBank mRNA BC136994.1 - 1 795
BC136995.1 - 1 795 9020987

Gene Symbol
OR2T12
Entrez Gene ID
127064
Gene Name
olfactory receptor family 2 subfamily T member 12
Gene Aliases
OR1-57
Chromosome Location
Chr. 1: 248294616 - 248295578 on Build GRCh38
UniGene ID
Hs.553582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004692.1 NP_001004692.1 1 773

Gene Symbol
OR2T8
Entrez Gene ID
343172
Gene Name
olfactory receptor family 2 subfamily T member 8
Gene Aliases
OR2T8P
Chromosome Location
Chr. 1: 247914138 - 247923152 on Build GRCh38
UniGene ID
Hs.521114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005522.1 NP_001005522.1 1 773
XM_011544178.2 XP_011542480.1 2 2116
XM_017001172.1 XP_016856661.1 2 4895

View Full Product Details