Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 8 | 13618 | |
NM_033026.5 | NP_149015.2 | 8 | 13618 | ||
XM_017012006.1 | XP_016867495.1 | 6 | 7798 | ||
XM_017012007.1 | XP_016867496.1 | 6 | 7782 | ||
GenBank mRNA | AB011131.1 | - | 2 | 2162 |