Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161683.1 | NP_001155155.1 | 4 | 566 | |
NM_144672.3 | NP_653273.3 | 8 | 710 | ||
XM_011545747.2 | XP_011544049.1 | 9 | 1089 | ||
XM_017022950.1 | XP_016878439.1 | 9 | 813 | ||
GenBank mRNA | AK125840.1 | - | 4 | 566 | |
BC129992.1 | - | 8 | 707 | ||
BC129993.1 | - | 8 | 714 | 40128985 |