Gene Transcripts
- Gene Symbol
- OTOA
- Entrez Gene ID
- 146183
- Gene Name
- otoancorin
- Gene Aliases
- CT108, DFNB22
- Chromosome Location
- Chr. 16: 21663857 - 21771468 on Build GRCh38
- UniGene ID
- Hs.408336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161683.1 |
NP_001155155.1
|
12, 13 |
1663 |
|
NM_144672.3 |
NP_653273.3
|
16, 17 |
1807 |
|
NM_170664.2 |
NP_733764.1
|
7, 8 |
901 |
|
XM_011545747.2 |
XP_011544049.1
|
17, 18 |
2186 |
|
XM_011545748.2 |
XP_011544050.2
|
6, 7 |
1180 |
|
XM_017022950.1 |
XP_016878439.1
|
17, 18 |
1910 |
|
XM_017022951.1 |
XP_016878440.1
|
2, 3 |
204 |
|
GenBank mRNA |
AK093062.1 |
- |
7, 8 |
901 |
|
AK125840.1 |
- |
12, 13 |
1663 |
|
AK302733.1 |
- |
6, 7 |
963 |
|
BC129992.1 |
- |
16, 17 |
1804 |
|
BC129993.1 |
- |
16, 17 |
1811 |
40128985 |
View Full Product Details