Gene Transcripts

Gene Symbol
ADCY1
Entrez Gene ID
107
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, DFNB44
Chromosome Location
Chr. 7: 45574140 - 45723116 on Build GRCh38
UniGene ID
Hs.192215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281768.1 NP_001268697.1 9, 10 1302
XM_005249585.2 XP_005249642.1 8, 9 1614
GenBank mRNA AF086306.1 - 2, 3 218

View Full Product Details