Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000337.5 | NP_000328.2 | 5 | 859 | |
NM_001128209.1 | NP_001121681.1 | 4 | 813 | ||
NM_172244.2 | NP_758447.1 | 5 | 859 | ||
XM_005265966.4 | XP_005266023.1 | 5 | 687 | ||
XM_005265967.2 | XP_005266024.1 | 5 | 451 | ||
XM_011534621.2 | XP_011532923.1 | 4 | 516 | ||
XM_017009723.1 | XP_016865212.1 | 6 | 625 | ||
XM_017009724.1 | XP_016865213.1 | 6 | 902 | ||
GenBank mRNA | AK292794.1 | - | 5 | 451 | |
BC008431.1 | - | 7 | 758 | ||
BC020740.1 | - | 5 | 452 | 4702959 | |
BX537948.1 | - | 4 | 824 | ||
DA561069.1 | - | 4 | 406 | ||
U58331.1 | - | 4 | 378 | ||
X95191.1 | - | 3 | 340 |